Eugenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:97-53-0
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00008654
:164.2 g/mol
Merck Index:13,03931
Boiling Pt:255 °C (1013 hPa)
Melting Pt:∼–10 °C
Density:1.0652 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:110 °C
EINECS:202-589-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:2.46 g/l (25 °C)
- Melting Point:-9.2 °C
- Molar Mass:164.2 g/mol
- Boiling Point:252 °C (1013 hPa)
- Vapor Pressure:- 0.1 hPa (25 °C)
- Flash Point:110 °C
- Refractive Index:1.5405 (20 °C, 589 nm)
- Density:1.07 g/cm3 (20 °C)