Order Entry
Germany
ContactUsLinkComponent
Pyrocatechol
Pyrocatechol
Catalog # EHERC11060000
Supplier:  EHRENSTORFER
CAS Number:  
Pyrocatechol
Catalog # EHERC11060000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :110.11 g/mol
Merck Index:13,08089
Boiling Pt:245 °C (1013 hPa)
Melting Pt:103…105 °C
Density:1.0548 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-427-5
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:120-80-9
Formula:C₆H₆O₂
MDL Number:MFCD00002188