Pyrocatechol ≥99%
About this item
Purity: 99% CAS No.: 120-80-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 127°(260°F) Physical Form: Powder or flakes Density: 1.370 Sensitivity: Air and Light Sensitive Appearance: White to brown Odour: Aromatic Melting Point: 104-107° Boiling Point: 245-246° Beilstein Reference: 471401 Merck Reference: 14,7999 UN No.: 2811 MDL No.: MFCD00002188
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:14,7999
- Boiling Pt:245…246 °C
- Melting Pt:104…107 °C
- Density:1.370
- Flash Pt:127 °C (260 °F)
- Storage Temperature:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
4 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 120-80-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 127°(260°F) Physical Form: Powder or flakes Density: 1.370 Sensitivity: Air and Light Sensitive Appearance: White to brown Odour: Aromatic Melting Point: 104-107° Boiling Point: 245-246° Beilstein Reference: 471401 Merck Reference: 14,7999 UN No.: 2811 MDL No.: MFCD00002188
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:14,7999
- Boiling Pt:245…246 °C
- Melting Pt:104…107 °C
- Density:1.370
- Flash Pt:127 °C (260 °F)
- Storage Temperature:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



