
4-Methoxythiophenol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:696-63-9
:
Formula:C₇H₈OS
MDL Number:MFCD00004849
:140.21 g/mol
Boiling Pt:100…103 °C (13 mmHg)
Melting Pt:42 °C
Density:1.140
Flash Pt:96 °C (204 °F)
Storage Temperature:Ambient
EINECS:211-799-2
UN:2810
ADR:6.1,III
:GHS07