
4-Methoxythiophenol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:696-63-9
:
Formula:C₇H₈OS
MDL Number:MFCD00004849
:140.21 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:42 °C
Density:1.14 g/cm³
Flash Pt:96 °C
EINECS:211-799-2
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5820 to 1.5850 (20°C, 589 nm)
- GC:≥97.5 %
- TLC:≥95 %
- Appearance of solution:(0.5mmol/ml DMF) clear, colourless
- Specific optical rotation:+9.0° to +11.0° (20°C, 589 nm) (c=1 in MeOH)