


Diphenylamine ≥98% (by GC), puriss. p.a. ACS, reag. Ph.Eur., Sigma-Aldrich®
Catalog # SIAL33149-100G-R
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:169.23 g/mol
Merck Index:13,03349
Boiling Pt:302 °C (1013 hPa)
Melting Pt:53…54 °C
Density:1.165 g/cm³ (20 °C)
Flash Pt:153 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-539-4
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS06
:GHS08
:GHS09
CAS Number:122-39-4
Formula:(C₆H₅)₂NH
MDL Number:MFCD00003014