Diphenylamine 99+%, extra pure indicator grade, redox indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-39-4
:
Formula:(C₆H₅)₂NH
MDL Number:MFCD00003014
:169.23 g/mol
Merck Index:13,03349
Boiling Pt:302 °C (1013 hPa)
Melting Pt:52…54 °C
Density:1.165 g/cm³ (20 °C)
Flash Pt:152 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-539-4
UN:3077
ADR:9,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Sulfated ash:≤0.05 %
- GC:≥99.0 %
- Sensitivity:(to nitrate) Passes test