Sulphanilamide ≥98%, TECHNICAL
Plastic bottle for solids, 250 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: 4-Aminobenzenesulphonamide
- CAS Number:63-74-1
- Formula:H₂NC₆H₄SO₂NH₂
- MDL Number:MFCD00007939
- Molecular Weight:172.21 g/mol
- Merck Index:13,09010
- Boiling Pt:401 °C (1013 hPa)
- Melting Pt:163…167 °C
- Density:1.46 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-563-4
Documents
Certificates
Search by Lot Number750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
P20_VWRC21156.237_SPEC_EN.pdf spec_VWRC21156.237 spec_VWRC21156.237 spec_VWRC21156.237
Product specification
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g



