Order Entry
Germany
ContactUsLinkComponent
3-Aminophenol
3-Aminophenol
Catalog # PROOMM0042.07
CAS Number:  
3-Aminophenol
Catalog # PROOMM0042.07
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Merck Index:13,00459
Boiling Pt:299 °C (1013 hPa)
Melting Pt:122 °C
Density:1.275 g/cm³ (20 °C)
Flash Pt:92 °C
EINECS:209-711-2
UN:2512
ADR:6.1,III
 :109.13 g/mol
 :GHS07
 :GHS09
CAS Number:591-27-5
Formula:C₆H₇NO
MDL Number:MFCD00007786