2-Amino-5-nitrophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121-88-0
:
Formula:C₆H₆N₂O₃
MDL Number:MFCD00007692
:154.13 g/mol
Melting Pt:207…208 °C
Density:1.54 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:204-503-8
:GHS07