2-Amino-5-nitrophenol
Catalog # MOLE24240078-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:154.13 g/mol
Melting Pt:207…208 °C
Density:1.54 g/cm³ (20 °C)
EINECS:204-503-8
:GHS07
CAS Number:121-88-0
Formula:C₆H₆N₂O₃
MDL Number:MFCD00007692