2-Amino-5-nitrophenol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121-88-0
:
Formula:C₆H₆N₂O₃
MDL Number:MFCD00007692
:154.13 g/mol
Melting Pt:∼ 200 °C (dec.)
Density:1.54 g/cm³ (20 °C)
EINECS:204-503-8
:GHS08
:GHS07
Storage Temperature:Ambient