


Phenyl isothiocyanate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-72-0
:
Formula:C₇H₅NS
MDL Number:MFCD00004798
:135.19 g/mol
Merck Index:12,07450
Boiling Pt:221 °C (760 mmHg)
Melting Pt:–21 °C
Density:1.1288 g/cm³
Flash Pt:87 °C
Storage Temperature:Refrigerator
EINECS:203-138-1
UN:2927
ADR:6.1,II
:GHS06
:GHS08
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6490 to 1.6510 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥97.5 %
- Specific gravity:(20°C) 1.13 to 1.136