
3-Methoxyphenyl isocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18908-07-1
:
Formula:C₈H₇NO₂
MDL Number:MFCD00002019
:149.15 g/mol
Boiling Pt:94…95 °C (10 mmHg)
Density:1.13 g/cm³
Flash Pt:96 °C
EINECS:242-658-3
UN:2206
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Clear colorless to light pink Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5420 to 1.5440 (20°C, 589 nm)
- GC:≥98.5 %