
4-Iodobenzenesulphonyl chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-61-3
:
Formula:C₆H₄ClIO₂S
MDL Number:MFCD00007441
:302.52 g/mol
Melting Pt:80…84 °C
EINECS:202-686-9
UN:1759
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to beige to orange Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:80 to 84 °C
- Titration Argentometric:≥96.0 % (ex Cl)
- GC:≥96.0 %