Order Entry
Germany
ContactUsLinkComponent
Methyl-2-iodobenzoate 99+%
Irritant
Methyl-2-iodobenzoate 99+%
Certificates
undefined
Irritant
Methyl-2-iodobenzoate 99+%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6020 to 1.6060 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together