4-Fluoroanisole, Sigma-Aldrich®
SIALF4602-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:126.13 g/mol
:157 °C (1013 hPa)
:–43.5 °C
:1.1178 g/cm³ (18.3 °C)
:207-295-7
:3271
:3,III
:M-Clarity™ quality level = MQ100
:459-60-9
:C₇H₇FO
:MFCD00000348