4-Fluorobenzoic acid, Sigma-Aldrich®
: Merck
:456-22-4
:
:C₇H₅FO₂
:MFCD00002530
:140.11 g/mol
:13,04198
:254 °C (1013 hPa)
:185 °C
:1.49 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:>100 °C
:207-259-0
:GHS07
:Ambient
Frequently Bought Together