Überspringen zu Kopf-Ebene
Überspringen zu Hauptinhalt
Überspringen zu Fußzeile
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluorobenzoic acid
2-Fluorobenzoic acid
2-Fluorobenzoic acid
CHMPFL20125.POR
:
CHEMPUR
:
2-Fluorobenzoic acid
CHMPFL20125.POR
:
CHEMPUR
:
Pk:
1
:
:
140.11 g/mol
:
245 °C (1013 hPa)
:
121…124 °C
:
1.46 g/cm³ (20 °C)
:
Ambient
:
207-158-1
:
GHS07
:
445-29-4
:
C₇H₅FO₂
:
MFCD00002405