6-Fluoroveratraldehyde
1
Catalog # CHMPFL13384.POR
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
- CAS Number:71924-62-4
- Formula:C₉H₉FO₃
- MDL Number:MFCD00061108
- Molecular Weight:184.17 g/mol
Specifications
Specifications
- Pk:1



