
Phenyl salicylate
Catalog # PROONE9304
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:214.22 g/mol
Merck Index:12,07464
Boiling Pt:307 °C (1013 hPa)
Melting Pt:41…43 °C
Density:1.307 g/cm³ (20 °C)
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:204-259-2
UN:3077
ADR:9,III
:GHS07
CAS Number:118-55-8
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002213