Phenyl salicylate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:118-55-8
:
Formula:C₁₃H₁₀O₃
MDL Number:MFCD00002213
:214.22 g/mol
Merck Index:12,07464
Boiling Pt:172…173 °C (12 mmHg)
Melting Pt:40…44 °C
Density:1,307 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:204-259-2
ADR:9,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:40 to 44 °C
- GC:≥98.5 %