ContactUsLinkComponent
3,4-Dimethoxybenzoic acid, Sigma-Aldrich®
  SIALD131806-100G
 :  Merck
 :  
3,4-Dimethoxybenzoic acid, Sigma-Aldrich®
  SIALD131806-100G
 :  Merck
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
 :
 :182.18 g/mol
 :12,10086
 :303 °C (1013 hPa)
 :117…119 °C
 :1.436 g/cm³ (–100 °C)
 :M-Clarity™ quality level = MQ200
 :>100 °C
 :Ambient
 :202-215-7
 :93-07-2
 :C₉H₁₀O₄
 :MFCD00002500