3,4-Dimethoxybenzoic acid
Catalog # PROOMM0319.01
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:182.18 g/mol
Merck Index:12,10086
Boiling Pt:303 °C (1013 hPa)
Melting Pt:117…119 °C
Density:1.436 g/cm³ (–100 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:202-215-7
CAS Number:93-07-2
Formula:C₉H₁₀O₄
MDL Number:MFCD00002500