3-Ethoxybenzoic acid
Glass bottle for solids, 1 g
Catalog # FLUO014643-1G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:621-51-2
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00016517
- Molecular Weight:166.18 g/mol
- EINECS:210-690-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



