
4-Cyanobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:619-65-8
:
Formula:C₈H₅NO₂
MDL Number:MFCD00002528
:147.13 g/mol
Boiling Pt:313 °C (1013 hPa)
Melting Pt:220…222 °C
Density:1.382 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:210-606-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together