Order Entry
Germany
ContactUsLinkComponent
Bis(2-ethylhexyl)phthalate
Bis(2-ethylhexyl)phthalate
Catalog # OVERDOP-25KG
CAS Number:  
undefined
Bis(2-ethylhexyl)phthalate
Catalog # OVERDOP-25KG
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :390.56 g/mol
Merck Index:13,02884
Boiling Pt:386.9 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:195 °C
Storage Temperature:Ambient
EINECS:204-211-0
UN:3082
ADR:9,III
 :GHS08
 :GHS07
CAS Number:117-81-7
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00009493