Order Entry
Germany
ContactUsLinkComponent
Butyl-4-hydroxybenzoate
Butyl-4-hydroxybenzoate
Catalog # PROOMM0496.00
CAS Number:  
Butyl-4-hydroxybenzoate
Catalog # PROOMM0496.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :194.23 g/mol
Merck Index:13,01583
Boiling Pt:156…157 °C (3.5 mmHg)
Melting Pt:73 °C
Density:1.23 g/cm³ (20 °C)
EINECS:202-318-7
CAS Number:94-26-8
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00016478