Butyl-4-hydroxybenzoate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-26-8
:
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00016478
:194.23 g/mol
Merck Index:13,01583
Boiling Pt:156 °C (3.5 mmHg)
Melting Pt:68…71 °C
Density:1,23 g/cm³ (20 °C)
Flash Pt:181 °C
EINECS:202-318-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:68 to 71 °C
- GC:≥99.0 %