Order Entry
Germany
ContactUsLinkComponent
Methyl-3-aminobenzoate
Methyl-3-aminobenzoate
Catalog # MAYBTL00154.50
Supplier:  Thermo Scientific
CAS Number:  
Methyl-3-aminobenzoate
Catalog # MAYBTL00154.50
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:120 °C (0.7 mmHg)
Melting Pt:148…152 °C
Density:1.232 g/cm³ (20 °C)
EINECS:224-842-5
CAS Number:4518-10-9
Formula:C₈H₉NO₂
MDL Number:MFCD00017102