
Methyl anthranilate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:134-20-3
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007710
:151.16 g/mol
Merck Index:14,6020
Boiling Pt:260 °C
Melting Pt:24 °C
Density:1.170
Flash Pt:123 °C (253 °F)
EINECS:205-132-4
:GHS07
Storage Temperature:Ambient