Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxybenzhydrazide

4-Hydroxybenzhydrazide

Glass bottle for solids, 100 g
Catalog # MOLE52538646-100G
Irritant
Marketsource
Supplier:   Molekula
4-Hydroxybenzhydrazide
Catalog # MOLE52538646-100G
Supplier:  Molekula
Marketsource
CAS Number:  5351-23-5
4-Hydroxybenzhydrazide

Glass bottle for solids, 100 g

CAS Number:  5351-23-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
  • CAS Number:5351-23-5
  • Formula:C₇H₈N₂O₂
  • MDL Number:MFCD00007605
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:370 °C (1013 hPa)
  • Melting Pt:266…269 °C
  • Storage Temperature:Ambient
  • EINECS:226-326-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications