Order Entry
Germany
ContactUsLinkComponent
3-Aminobenzamide
3-Aminobenzamide
Catalog # TREV4667-50-03
Supplier:  TREVIGEN
CAS Number:  
3-Aminobenzamide
Catalog # TREV4667-50-03
Supplier:  TREVIGEN
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :136.15 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:111…115 °C
EINECS:222-586-9
CAS Number:3544-24-9
Formula:C₇H₈N₂O
MDL Number:MFCD00007989