3-Aminobenzamide
Catalog # FLUO018167-5G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:136.15 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:111…115 °C
EINECS:222-586-9
CAS Number:3544-24-9
Formula:C₇H₈N₂O
MDL Number:MFCD00007989