Order Entry
Germany
ContactUsLinkComponent
3-Aminobenzamide, Millipore®
3-Aminobenzamide, Millipore®
Catalog # 165350-100
CAS Number:  
3-Aminobenzamide, Millipore®
Catalog # 165350-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :136.15 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:111…115 °C
EINECS:222-586-9
CAS Number:3544-24-9
Formula:C₇H₈N₂O
MDL Number:MFCD00007989