

Benzamide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55-21-0
:
Formula:C₇H₇NO
MDL Number:MFCD00007968
:121.14 g/mol
Merck Index:13,01059
Boiling Pt:228 °C (760 mmHg)
Melting Pt:125…129 °C
Density:1,35 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:200-227-7
:GHS07
:GHS08
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:125 to 129 °C
- HPLC:≥98.5 %
- UV:at 280 nm A: ≤0.05
- Titration with HClO4:99.4 to 100.6 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Calcium (Ca):≤0.001 %
- Iron (Fe):≤0.0005 %
- Sodium (Na):≤0.02 %
- Heavy metals (as Pb):≤5 ppm