3-Ethoxybenzonitrile 95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 95% CAS No.: 25117-75-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 110°(230°F) Refractive Index: 1.529 Boiling Point: 241-244° UN No.: 3276 MDL No.: MFCD00001805
- CAS Number:25117-75-3
- Formula:C₉H₉NO
- MDL Number:MFCD00001805
- Molecular Weight:147.18 g/mol
- Boiling Pt:241…244 °C
- Density:1.047
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:246-632-2
- UN:3276
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 25117-75-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 110°(230°F) Refractive Index: 1.529 Boiling Point: 241-244° UN No.: 3276 MDL No.: MFCD00001805
- CAS Number:25117-75-3
- Formula:C₉H₉NO
- MDL Number:MFCD00001805
- Molecular Weight:147.18 g/mol
- Boiling Pt:241…244 °C
- Density:1.047
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:246-632-2
- UN:3276
- ADR:6.1,III
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



