2-Chloro-4-fluorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL297836-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:174.56 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:218-845-0
CAS Number:2252-51-9
Formula:C₇H₄ClFO₂
MDL Number:MFCD00010615