Order Entry
Germany
ContactUsLinkComponent
 
Affinity chromatography columns, HiScreen™ MabSelect™ PrismA
  17-5498-15
 :  Cytiva
undefined
Affinity chromatography columns, HiScreen™ MabSelect™ PrismA
  17-5498-15
 :  Cytiva

 

  • Particle Size:
    60 µm
  • Max. pressure:
    3 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ MabSelect™ PrismA
  • Seq:
    01
  • pH range:
    3 – 12
  • Temperature limit:
    40 °C
  • Binding capacity:
    ~ 80 mg human IgG/ml resin at 6 min residence time
    ~ 40 mg human IgG/ml resin at 2 min residence time
  • Storage:
    20% ethanol, 2 to 8 °C
  • Column hardware:
    PP
  • Bed dimensions:
    7,7×100 mm
  • Flow velocity:
    <500 cm/h
  • Pk:
    1 mL

 

 

These columns are prepacked with MabSelect™ PrismA protein A chromatographyn resin that has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs. These columns are suitable for purifying antibodies with affinity chromatography in research and process development.

  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH
  • Convenient HiScreen™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
  • Convenient method optimisation and parameter screening enabled by the 10 cm bed height

MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.

The columns are made of biocompatible PP that does not interact with biomolecules.

 : Note that HiScreen™ columns cannot be opened or repacked.