3,4,5-Trimethoxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:86-81-7
:
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00003364
:196.2 g/mol
Boiling Pt:311 °C (1013 hPa)
Melting Pt:72…75 °C
Density:1.367 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-701-6