3-Hydroxy-p-anisaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:621-59-0
:
Formula:C₈H₈O₃
MDL Number:MFCD00003369
:152.15 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.401 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:210-694-9