Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dibromo-4-fluoroaniline
2,6-Dibromo-4-fluoroaniline
2,6-Dibromo-4-fluoroaniline
2,6-Dibromo-4-fluoroaniline
Supplier:
FLUOROCHEM
Certificates
Expand image
2,6-Dibromo-4-fluoroaniline
Supplier:
FLUOROCHEM
About this item
CAS Number:
344-18-3
:
Formula:
C₆H₄Br₂FN
MDL Number:
MFCD00041445
:
268.91 g/mol
Boiling Pt:
277 °C (1013 hPa)
Melting Pt:
68 °C
UN:
2811
ADR:
6.1,III