1,3-Dibromotetrafluorobenzene, Sigma-Aldrich®
Catalog # SIAL102016-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:307.87 g/mol
Boiling Pt:85 °C (18 mmHg)
:M-Clarity™ quality level = MQ100
EINECS:216-326-3
:GHS07
CAS Number:1559-87-1
Formula:C₆Br₂F₄
MDL Number:MFCD00031533