Methyl-3-bromobenzoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:618-89-3
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00017777
:215.05 g/mol
Boiling Pt:69…70 °C (0.2 mmHg)
Melting Pt:31 °C
:M-Clarity™ quality level = MQ200
EINECS:210-569-9
Frequently Bought Together