Order Entry
Germany
ContactUsLinkComponent
pH Indicator solution, full range pH 1-14
pH Indicator solution, full range pH 1-14
Catalog # DIUKUN10900-135
Supplier:  DISTRIBUTOR
CAS Number:  
pH Indicator solution, full range pH 1-14
Catalog # DIUKUN10900-135
Supplier:  DISTRIBUTOR
CAS Number:  

About this item

 :