Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromophenylboronic acid
2-Bromophenylboronic acid ≥98%
2-Bromophenylboronic acid ≥98%
:
Thermo Fisher Scientific
2-Bromophenylboronic acid ≥98%
:
Thermo Fisher Scientific
:
244205-40-1
:
:
C₆H₆BBrO₂
:
MFCD01114672
:
200.83 g/mol
:
108…110 °C
:
GHS07
:
Ambient
msds_ALFAL18581
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years