
3-Methoxyphenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10365-98-7
:
Formula:C₇H₉BO₃
MDL Number:MFCD00161359
:151.96 g/mol
Melting Pt:149…154 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %