Dichlone
Catalog # MOLEM50997691
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:227.05 g/mol
Merck Index:13,03071
Boiling Pt:304 °C (1013 hPa)
Melting Pt:193 °C
Density:1.44 g/cm³ (16 °C)
EINECS:204-210-5
UN:2811
ADR:6.1,III
:GHS07
:GHS09
CAS Number:117-80-6
Formula:C₁₀H₄Cl₂O₂
MDL Number:MFCD00001677