Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Isopropylisatin
1-Isopropylisatin
1-Isopropylisatin
1-Isopropylisatin
Supplier:
BIONET RESEARCH
Certificates
Expand image
1-Isopropylisatin
Supplier:
BIONET RESEARCH
About this item
CAS Number:
10487-31-7
:
Formula:
C₁₁H₁₁NO₂
MDL Number:
MFCD00224231
:
189.21 g/mol