0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
7-Chloroisatin
7-Chloroisatin, Sigma-Aldrich®
7-Chloroisatin, Sigma-Aldrich®
SIAL675040-1G
:
Merck
:
7-Chloroisatin, Sigma-Aldrich®
SIAL675040-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
181.58 g/mol
:
GHS07
:
7477-63-6
:
C₈H₄ClNO₂
:
MFCD00022796